Skip to Content
Merck
All Photos(2)

Documents

685380

Sigma-Aldrich

Carbon nanotube, single-walled

amide functionalized, >90% carbon basis, D × L4-6 nm × 0.7-1.0 μm , bundle dimensions

Synonym(s):

SWNT, amide functionalized

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

product name

Carbon nanotube, single-walled, amide functionalized, >90% carbon basis, D × L 4-6 nm × 0.7-1.0 μm , bundle dimensions

Assay

>90% carbon basis

form

powder

extent of labeling

per 4 atom % (amide groups)

D × L

4-6 nm × 0.7-1.0 μm , bundle dimensions

impurities

5-8% metals

solubility

DMF: 0.5-1.0 mg/mL (with sonication)
acetone: 0.5-1.0 mg/mL (with sonication)
alcohols: 0.5-1.0 mg/mL (with sonication)

functional group

amide

Preparation Note

Electric Arc Discharge Method

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular characterization of toxicity mechanism of single-walled carbon nanotubes.
Chen PH, et al.
Biomaterials, 34(22), 5661-5669 (2013)
Vibration characteristics of single- and double-walled carbon nanotubes functionalized with amide and amine groups
Ajori R, et al.
Physica B: Condensed Matter, 462, 8-1 (2015)

Articles

Carbon nanotubes are materials that possess remarkable properties and offer extraordinary possibilities.

Carbon nanotubes (CNTs) have received much attention since their discovery in 1991 by Sumio lijima1 due to their excellent mechanical, electrical, and optical properties.

A nanocomposite is typically defined as a mixture between a host material (e.g., polymer matrix) and nanofillers with at least one dimension of less than 100 nm.

SWCNTs show promise in FETs, solar cells, and photodetectors due to their ultrafast charge transport mobility.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service