Skip to Content
Merck
All Photos(1)

Key Documents

682527

Sigma-Aldrich

6-(Ferrocenyl)hexanethiol

Synonym(s):

FHT, 6-(Mercaptohexyl)ferrocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22FeS
CAS Number:
Molecular Weight:
302.26
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

bp

321-353 °C

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.SCCCCCC[C]2[CH][CH][CH][CH]2

InChI

1S/C11H17S.C5H5.Fe/c12-10-6-2-1-3-7-11-8-4-5-9-11;1-2-4-5-3-1;/h4-5,8-9,12H,1-3,6-7,10H2;1-5H;

InChI key

YENJERDZPAXYRX-UHFFFAOYSA-N

General description

6-(Ferrocenyl)hexanethiol (FHT) is a ferrocenyl alkanethiol that forms a self-assembled monolayer (SAM) where ferrocenyl linkages can facilitate the electron exchange between the coating and the substrate.[1]

Application

FHT can be used as a SAM on gold substrates to act as an electron transport medium which can be potentially used in bio-electrochemical devices.[2] FHT may also be used to functionalize graphene coated gold surfaces by forming gold-thiol linkages which can be used as electron transfer mediators.[3]
Monolayers of this material are easy to fabricate and provide a stable and reproducible system and are used to detect molecular interactions.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N H Somé et al.
Social science & medicine (1982), 264, 113310-113310 (2020-09-03)
Team-based primary care offers a wide range of health services to patients by using interdisciplinary health care providers committed to delivering comprehensive, coordinated and high-quality care through team collaboration. Ontario's Family Health Team (FHT), the largest team-based practice model in
Man Yi Ho et al.
The journal of physical chemistry. B, 114(32), 10661-10665 (2010-08-13)
Ferrocene-terminated self-assembled monolayers (Fc-SAMs) are one of the most studied molecular aggregates on metal electrodes. They are easy to fabricate and provide a stable and reproducible system to investigate the effect of the microenvironment on the electron transfer parameters. We
Xiaoli Wang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 985, 14-28 (2015-02-07)
Fangji Huangqi Tang (FHT) is a classical formula widely used in Chinese clinical application. In this paper, a novel and advanced strategy has been developed for the multiple constituent identification of FHT in rats, which was basing on an ultra-high
Fernando Silva-Oliveira et al.
International journal of paediatric dentistry, 29(5), 624-634 (2019-05-10)
Child maltreatment is a complex social and public health problem. To evaluate the frequency of the recognition by Brazilian health professionals in primary care of child physical abuse (CPA) and associated factors. A representative cross-sectional study was conducted with dentists
Hui Gu et al.
Biosensors & bioelectronics, 135, 111-119 (2019-04-21)
The detection of Cu2+ ion, one of the metal ions substantial in cerebral physiology, is critical in studying brain activities and understanding brain functions. However, repetitive measurements of Cu2+ in the progress of physiological and pathological events is still challenging

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service