Skip to Content
Merck
All Photos(3)

Key Documents

681334

Sigma-Aldrich

2-(Trimethylsilyl)ethanesulfonyl chloride

Synonym(s):

2-Trimethylsilylethylsulfonyl chloride, SES-Cl

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$459.00
5 G
$1,590.00

$459.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$459.00
5 G
$1,590.00

About This Item

Linear Formula:
(CH3)3SiCH2CH2SO2Cl
CAS Number:
Molecular Weight:
200.76
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$459.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

refractive index

n20/D 1.4444

bp

146.8 °C/760 mmHg

density

1.059 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C[Si](C)(C)CCS(Cl)(=O)=O

InChI

1S/C5H13ClO2SSi/c1-10(2,3)5-4-9(6,7)8/h4-5H2,1-3H3

InChI key

BLPMCIWDCRGIJV-UHFFFAOYSA-N

Application

Reactant involved in:
  • Nucleophilic substitution for synthesis of nicotinamine and its analogs
  • Regioselective metal-free oxidative cyclization of sulfonamides
  • Annulation reactions
  • Tin-free radical carbonylation of alkylsulfonyl derivatives
  • Asymmetric aziridination
Reagent used for protection of an amino group as a SES-amide. The SES group can be conveniently removed by fluoride ion.[1]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

70.0 °F - closed cup

Flash Point(C)

21.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-(Trimethylsilyl)ethanesulfonyl (or SES) group in amine protection and activation.
Patrice Ribière et al.
Chemical reviews, 106(6), 2249-2269 (2006-06-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service