Skip to Content
Merck
All Photos(2)

Key Documents

681326

Sigma-Aldrich

2-(Trimethylsilyl)ethanesulfonamide

90%

Synonym(s):

SES-NH2

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$342.00

$342.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$342.00

About This Item

Linear Formula:
(CH3)3SiCH2CH2SO2NH2
CAS Number:
Molecular Weight:
181.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$342.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

90%

form

solid

mp

81-87 °C

SMILES string

C[Si](C)(C)CCS(N)(=O)=O

InChI

1S/C5H15NO2SSi/c1-10(2,3)5-4-9(6,7)8/h4-5H2,1-3H3,(H2,6,7,8)

InChI key

MZASHBBAFBWNFL-UHFFFAOYSA-N

Application

Useful reagent for introduction of a SES-protected nitrogen functionality, which can be cleaved with fluoride ion.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-(Trimethylsilyl)ethanesulfonyl (or SES) group in amine protection and activation.
Patrice Ribière et al.
Chemical reviews, 106(6), 2249-2269 (2006-06-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service