Skip to Content
Merck
All Photos(1)

Key Documents

680710

Sigma-Aldrich

Rhodium on carbon

Evonik NOBLYST® P3053 5% Rh

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$556.00

$556.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$556.00

About This Item

Linear Formula:
Rh
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$556.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

reaction suitability

core: rhodium
reagent type: catalyst

extent of labeling

5 wt. % loading (dry basis)

weight

Unit excludes wt. of H2O

matrix

carbon, wet support

SMILES string

[Rh]

InChI

1S/Rh

InChI key

MHOVAHRLVXNVSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Legal Information

Noblyst is a registered trademark of Evonik Degussa GmbH

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

An application guide to help you select the most suitable heterogeneous catalysts for diverse synthetic applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service