Skip to Content
Merck
All Photos(1)

Key Documents

84031

Sigma-Aldrich

Ruthenium

5% Ru basis on activated charcoal

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$89.90

$89.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$89.90

About This Item

UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$89.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Ruthenium 5% on activated charcoal, 5% Ru basis

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

concentration

5% Ru

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service