Skip to Content
Merck
All Photos(1)

Key Documents

680044

Sigma-Aldrich

6-Chloropyridine-2-carboxylic acid

97%

Synonym(s):

6-Chloropicolinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClNO2
CAS Number:
Molecular Weight:
157.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

185-190 °C

SMILES string

OC(=O)c1cccc(Cl)n1

InChI

1S/C6H4ClNO2/c7-5-3-1-2-4(8-5)6(9)10/h1-3H,(H,9,10)

InChI key

ZLKMOIHCHCMSFW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charalambos Skoutelis et al.
Journal of hazardous materials, 321, 753-763 (2016-10-12)
2-Chloropyridine, an important precursor of the chemical industry is also a persistent water pollutant. The genotoxicity of photolytically treated 2-chloropyridine aqueous solution to human lymphocytes initially increases and fluctuates during treatment finally reaching the control values after prolonged treatment. Intermediate
[Determination of nitrapyrin and its metabolite 6-chloropicolinic acid in the environment].
Iu A Buniatian et al.
Gigiena i sanitariia, (4)(4), 52-53 (1983-04-01)
Analytical method for nitrapyrin and 6-chloropicolinic acid residues in strawberry fruit and soil.
Y Iwata et al.
Journal of agricultural and food chemistry, 29(2), 235-239 (1981-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service