Skip to Content
Merck
All Photos(1)

Key Documents

675946

Sigma-Aldrich

(S)-(−)-2,2′,6,6′-Tetramethoxy-4,4′-bis[di(3,5-xylyl)phosphino]-3,3′-bipyridine

97%

Synonym(s):

(S)-Xylyl-P-Phos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H50N2O4P2
CAS Number:
Molecular Weight:
756.85
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

158-162 °C

functional group

phosphine

SMILES string

COc1cc(P(c2cc(C)cc(C)c2)c3cc(C)cc(C)c3)c(c(OC)n1)-c4c(OC)nc(OC)cc4P(c5cc(C)cc(C)c5)c6cc(C)cc(C)c6

InChI

1S/C46H50N2O4P2/c1-27-13-28(2)18-35(17-27)53(36-19-29(3)14-30(4)20-36)39-25-41(49-9)47-45(51-11)43(39)44-40(26-42(50-10)48-46(44)52-12)54(37-21-31(5)15-32(6)22-37)38-23-33(7)16-34(8)24-38/h13-26H,1-12H3

InChI key

JRTHAKOHBMETRC-UHFFFAOYSA-N

Application

Ligand employed in the asymmetic hydrogenation of ß-keto esters, 2-arylacrylates, aryl ketones , and other substrates.[1]

Legal Information

Sold in collaboration with Johnson Matthey Catalyst for research purposes only. US5886182 and any patents arising therefrom apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Wu et al.
Accounts of chemical research, 39(10), 711-720 (2006-10-18)
This Account outlines our efforts in the design and synthesis of a family of highly effective atropisomeric dipyridylphosphine ligands (P-Phos and its variants) and in the development of their widespread applications in transition-metal-catalyzed asymmetric reactions including hydrogenation, hydrosilylation, and C-C

Articles

P-Phos ligand family enhances catalysis, featuring atropisomeric biaryl bisphosphine with unique structural elements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service