Skip to Content
Merck
All Photos(1)

Key Documents

675806

Sigma-Aldrich

(R)-(+)-2,2′,6,6′-Tetramethoxy-4,4′-bis(diphenylphosphino)-3,3′-bipyridine

97%

Synonym(s):

(R)-P-Phos

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$129.00

$129.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$129.00

About This Item

Empirical Formula (Hill Notation):
C38H34N2O4P2
CAS Number:
Molecular Weight:
644.64
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$129.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

optical activity

[α]20/D +98°, c = 1 in chloroform

mp

261-265 °C

functional group

phosphine

SMILES string

COc1cc(P(c2ccccc2)c3ccccc3)c(c(OC)n1)-c4c(OC)nc(OC)cc4P(c5ccccc5)c6ccccc6

InChI

1S/C38H34N2O4P2/c1-41-33-25-31(45(27-17-9-5-10-18-27)28-19-11-6-12-20-28)35(37(39-33)43-3)36-32(26-34(42-2)40-38(36)44-4)46(29-21-13-7-14-22-29)30-23-15-8-16-24-30/h5-26H,1-4H3

InChI key

JZOSBBLJKXSBBN-UHFFFAOYSA-N

Application

Ligand employed in the asymmetic hydrogenation of ß-keto esters, 2-arylacrylates, aryl ketones , and other substrates.[1]

Legal Information

Sold in collaboration with Johnson Matthey Catalyst for research purposes only. US5886182 and any patents arising therefrom apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Wu et al.
Accounts of chemical research, 39(10), 711-720 (2006-10-18)
This Account outlines our efforts in the design and synthesis of a family of highly effective atropisomeric dipyridylphosphine ligands (P-Phos and its variants) and in the development of their widespread applications in transition-metal-catalyzed asymmetric reactions including hydrogenation, hydrosilylation, and C-C

Articles

P-Phos ligand family enhances catalysis, featuring atropisomeric biaryl bisphosphine with unique structural elements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service