Skip to Content
Merck
All Photos(1)

Key Documents

692395

Sigma-Aldrich

(R)-SEGPHOS®

≥94%

Synonym(s):

(R)-(+)-5,5′-Bis(diphenylphosphino)-4,4′-bi-1,3-benzodioxole, [4(R)-(4,4′-bi-1,3-benzodioxole)-5,5′-diyl]bis[diphenylphosphine]

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$58.30

$58.30


Estimated to ship onMarch 21, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$58.30

About This Item

Empirical Formula (Hill Notation):
C38H28O4P2
CAS Number:
Molecular Weight:
610.57
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$58.30


Estimated to ship onMarch 21, 2025


Request a Bulk Order

Quality Level

Assay

≥94%

form

powder

optical activity

[α]20/D +11°, c = 0.5 in chloroform

mp

168-172 °C

functional group

phosphine

SMILES string

C1Oc2ccc(P(c3ccccc3)c4ccccc4)c(c2O1)-c5c6OCOc6ccc5P(c7ccccc7)c8ccccc8

InChI

1S/C38H28O4P2/c1-5-13-27(14-6-1)43(28-15-7-2-8-16-28)33-23-21-31-37(41-25-39-31)35(33)36-34(24-22-32-38(36)42-26-40-32)44(29-17-9-3-10-18-29)30-19-11-4-12-20-30/h1-24H,25-26H2

InChI key

RZZDRSHFIVOQAF-UHFFFAOYSA-N

Application

Catalytic ligand used for:
  • Nickel-catalyzed asymmetric α-arylation and heteroarylation of ketones with chloroarenes
  • Enantioselective synthesis of dihydrobenzofurans and dihydronaphthofurans via olefin isomerization/enantioselective intramolecular Alder-ene reaction of enynes catalyzed by Rh
  • Preparation of axially chiral biaryl compounds by gold-catalyzed stereoselective intramolecular hydroarylation
  • Preparation of chiral silylated homoallylic alcohols and diols by asymmetric addition of alcohols and aldehydes to silylbutadienes catalyzed by ruthenium complexes
  • Preparation of chiral 3-alkyl-substituted indolines by tandem condensation-asymmetric hydrogenation of indoles with aldehydes, catalyzed by Bronsted acids and palladium BINAP complexes
  • Rhodium-catalyzed asymmetric formal olefination or cycloaddition of 1,3-dicarbonyl compounds with 1,6-diynes or 1,6-enynes

Legal Information

Sold in collaboration with Takasago for research purposes only. JP Registration No. 3148136
SEGPHOS is a registered trademark of Takasago Intl. Corp.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We present an article concerning BINAP/SEGPHOS® Ligands and Complexes.

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service