Skip to Content
Merck
All Photos(1)

Key Documents

673722

Sigma-Aldrich

3-Chloro-2-hydroxybenzaldehyde

97%

Synonym(s):

3-Chlorosalicylaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$159.00

$159.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$159.00

About This Item

Empirical Formula (Hill Notation):
C7H5ClO2
CAS Number:
Molecular Weight:
156.57
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$159.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

97%

form

solid

mp

52-56 °C

functional group

aldehyde
chloro

SMILES string

[H]C(=O)c1cccc(Cl)c1O

InChI

1S/C7H5ClO2/c8-6-3-1-2-5(4-9)7(6)10/h1-4,10H

InChI key

DOHOPUBZLWVZMZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service