Skip to Content
Merck
All Photos(1)

Key Documents

261815

Sigma-Aldrich

3,5-Dichlorosalicylaldehyde

99%

Synonym(s):

2,4-Dichloro-6-formylphenol, 2-Hydroxy-3,5-dichlorobenzaldehyde, 3,5-Dichloro-2-hydroxybenzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$47.10
25 G
$153.00

$47.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$47.10
25 G
$153.00

About This Item

Linear Formula:
Cl2C6H2(OH)CHO
CAS Number:
Molecular Weight:
191.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$47.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

solid

mp

95-97 °C (lit.)

solubility

ethanol: soluble 5%, clear, faintly yellow to greenish-yellow

functional group

aldehyde
chloro

SMILES string

Oc1c(Cl)cc(Cl)cc1C=O

InChI

1S/C7H4Cl2O2/c8-5-1-4(3-10)7(11)6(9)2-5/h1-3,11H

InChI key

FABVMBDCVAJXMB-UHFFFAOYSA-N

Application

3,5-Dichlorosalicylaldehyde has been used in the preparation of:
  • Schiff base ligands[1]
  • (RS)-3,5-dichloro-2-[[(1-phenylethyl)imino]methyl]phenol[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashiro Akitsu et al.
Acta crystallographica. Section C, Crystal structure communications, 60(Pt 11), o801-o802 (2004-11-06)
The title compound, (RS)-3,5-dichloro-2-[[(1-phenylethyl)imino]methyl]phenol, C(15)H(13)Cl(2)NO, was synthesized from racemic 1-phenylethylamine and 3,5-dichlorosalicylaldehyde. The pi-conjugate system around the imine group is essentially planar in the phenol-imine tautomer. Intramolecular O...N hydrogen-bond and intermolecular C-H...pi interactions are present in the crystal structure.
Ishani Majumder et al.
Inorganic chemistry, 56(21), 12893-12901 (2017-10-24)
A tridentate N,N,O donor ligand 2,4-dichloro-2-[(2-piperazine-4-yl-ethylimino)-methyl]-phenol (HL) was designed, and eight new Zn
Yongming Cui et al.
European journal of medicinal chemistry, 58, 323-331 (2012-11-13)
Eleven mononuclear copper(II), nickel(II), zinc(II) and cobalt(II) complexes of Schiff base ligands derived from 3,5-dibromosalicylaldehyde/3,5-dichlorosalicylaldehyde were synthesized and determined by single crystal X-ray analysis. The crystal structures of complexes 1, 2, 4, 5, 6, 8 and 11 present the square-planar

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service