Skip to Content
Merck
All Photos(1)

Key Documents

673307

Sigma-Aldrich

Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate

Synonym(s):

(Cyclopentadienyltris(acetonitrile)ruthenium hexafluorophosphate, Tris(acetonitrile)cyclopentadienylruthenium hexafluorophosphate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$230.00
1 G
$524.00

$230.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$230.00
1 G
$524.00

About This Item

Linear Formula:
[(C5H5)Ru(CH3CN)3]PF6
CAS Number:
Molecular Weight:
434.28
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$230.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

reaction suitability

core: ruthenium
reagent type: catalyst

storage temp.

2-8°C

SMILES string

[Ru+].CC#N.CC#N.CC#N.[CH]1[CH][CH][CH][CH]1.F[P-](F)(F)(F)(F)F

InChI

1S/C5H5.3C2H3N.F6P.Ru/c1-2-4-5-3-1;3*1-2-3;1-7(2,3,4,5)6;/h1-5H;3*1H3;;/q;;;;-1;+1

InChI key

HJQVFVSAQOQXRG-UHFFFAOYSA-N

Application

Catalyst for:
  • Regioselective and stereoselective dehydrative allylation of nucleophiles
  • O-H Insertion and condensation reactions
  • Hydrosilylation
  • Decarboxylative allylic etherification
  • Claisen rearrangement of unactivated allyl vinyl ethers

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiuzhong Zhu et al.
Polymers, 11(4) (2019-04-10)
A series of side-chain functionalized polytetrahydrofuran (PTHF) derivatives were synthesized via the blue-light photocatalytic thiol-ene "click" reaction. Firstly, unsaturated polytetrahydrofuran (UPTHF) as a new unsaturated polyether was synthesized via condensation polymerization of cis-2-butene-1,4-diol and trans-1,4-dibromo-2-butene using potassium hydroxide (KOH) as

Related Content

The Herzon group has developed highly active catalysts for the anti-Markovnikov hydration and reductive hydration of terminal alkynes to form aldehydes and linear alcohols, respectively.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service