Skip to Content
Merck
All Photos(1)

Documents

673269

Sigma-Aldrich

Sodium dimethylphenylsilanolate hydrate

97%

Synonym(s):

Phenyl(dimethyl)silanolate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)Si(ONa)(CH3)2 · xH2O
Molecular Weight:
174.25 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

197-218 °C

SMILES string

[Na+].[H]O[H].C[Si](C)([O-])c1ccccc1

InChI

1S/C8H11OSi.Na.H2O/c1-10(2,9)8-6-4-3-5-7-8;;/h3-7H,1-2H3;;1H2/q-1;+1;

InChI key

GTSOGRDRMFDZQL-UHFFFAOYSA-N

Application

A useful silicon nucleophile for Pd-catalyzed cross-coupling.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denmark, S. E.; Ober, M. H.
Aldrichimica Acta, 36, 75-75 (2003)

Articles

Pd-catalyzed cross-coupling of silicon compounds offers an alternative in synthetic chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service