Skip to Content
Merck
All Photos(1)

Key Documents

394513

Sigma-Aldrich

Paraformaldehyde-d2

98 atom % D, 98% (CP)

Synonym(s):

Deuterated paraformaldehyde, Dideuteroparaformaldehyde, Formaldehyde-d2 polymer, Polyoxymethylene-D2

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$339.00
5 G
$1,080.00

$339.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$339.00
5 G
$1,080.00

About This Item

Linear Formula:
(CD2O)n
CAS Number:
Molecular Weight:
32.04
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

$339.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

98% (CP)

form

powder

mp

170 °C (dec.) (lit.)

mass shift

M+2

storage temp.

room temp

SMILES string

[2H]C([2H])=O

InChI

1S/CH2O/c1-2/h1H2/i1D2

InChI key

WSFSSNUMVMOOMR-DICFDUPASA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Flam. Sol. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service