Skip to Content
Merck
All Photos(1)

Key Documents

671010

Sigma-Aldrich

O-(2,4,6-Trimethylbenzenesulfonyl)hydroxylamine

≥90%

Synonym(s):

O-(Mesitylenesulfonyl)hydroxylamine, MSH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO3S
CAS Number:
Molecular Weight:
215.27
MDL number:
UNSPSC Code:
12352000

Assay

≥90%

SMILES string

Cc1cc(C)c(c(C)c1)S(=O)(=O)ON

Citation

Aminating Reagent[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O-Mesitylenesulfonylhydroxylamine and Related Compounds - Powerful Aminating Reagents
Tamura, Y., et al.
Synthesis, 1-17 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service