Skip to Content
Merck
All Photos(1)

Key Documents

663557

Sigma-Aldrich

2-Amino-5-(4-fluorophenyl)-1,3,4-thiadiazole

97%

Synonym(s):

5-(4-Fluorophenyl)-1,3,4-thiadiazol-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6FN3S
CAS Number:
Molecular Weight:
195.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

238-243 °C

SMILES string

Nc1nnc(s1)-c2ccc(F)cc2

InChI

1S/C8H6FN3S/c9-6-3-1-5(2-4-6)7-11-12-8(10)13-7/h1-4H,(H2,10,12)

InChI key

WRSVCKNLHZWSNJ-UHFFFAOYSA-N

Application

2-Amino-5-(4-fluorophenyl)-1,3,4-thiadiazole can be used as a substrate in the preparation of:
  • Bithiophene based azo dyes with possible nonlinear optical (NLO) properties.
  • 4-Thiazolidinone derivatives as potent HCV NS5B polymerase inhibitors.
  • 1,3,4-Thiadiazole and phenothiazine hybrids as possible antitubercular agents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-Heteroarylimino-5-arylidene-4-thiazolidinones as a new class of non-nucleoside inhibitors of HCV NS5B polymerase
Kucukguzel ?, et al.
European Journal of Medicinal Chemistry, 69(2), 931-941 (2013)
Design of new phenothiazine-thiadiazole hybrids via molecular hybridization approach for the development of potent antitubercular agents
Ramprasad J, et al.
European Journal of Medicinal Chemistry, 106(2), 75-84 (2015)
Design, synthesis and evaluation of redox, second order nonlinear optical properties and theoretical DFT studies of novel bithiophene azo dyes functionalized with thiadiazole acceptor groups
Castro MR, et al.
Dyes and Pigments, 95(2), 392-399 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service