Skip to Content
Merck
All Photos(1)

Documents

438537

Sigma-Aldrich

2-Amino-5-phenyl-1,3,4-thiadiazole

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N3S
CAS Number:
Molecular Weight:
177.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

223-227 °C (lit.)

SMILES string

Nc1nnc(s1)-c2ccccc2

InChI

1S/C8H7N3S/c9-8-11-10-7(12-8)6-4-2-1-3-5-6/h1-5H,(H2,9,11)

InChI key

UHZHEOAEJRHUBW-UHFFFAOYSA-N

General description

2-Amino-5-phenyl-1,3,4-thiadiazole on condensation with benzaldehyde (SPT), 4-nitrobenzaldehyde (SNT), 4-methoxybenzaldehyde (SMT), 2-hydroxybenzaldehyde (SSTH) or 2-hydroxyacetophenone affords Schiff′s bases. Its molecular geometry and vibrational frequencies have been evaluated using the Hartree-Fock and density functional method (B3LYP). 2-Amino-5-phenyl-1,3,4-thiadiazole has been reported to inhibit the corrosion of mild steel in 0.5M H2SO4 and 1.0M HCl. Corrosion inhibition has been examined using potentiodynamic polarization and electrochemical impedance spectroscopy (EIS).

Application

2-Amino-5-phenyl-1,3,4-thiadiazole may be used for the synthesis of 1,3,4-thiadiazole derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization and antimicrobial activity of some carbamothioyl-1, 3, 4-thiadiazole derivatives.
Salih N, et al.
Synthesis, 4(2), 655-660 (2012)
A K Srivastava et al.
Bioinorganic chemistry and applications, 3(3-4), 289-297 (2008-03-28)
The reactions of bis(cyclopentadienyl)titanium(IV) dichloride with Schiff bases derived by condensing 2- amino-5-phenyl-1,3,4-thiadiazole with benzaldehyde (SPT), 4-nitrobenzaldehyde (SNT), 4-methoxybenzaldehyde (SMT), 2-hydroxybenzaldehyde (SSTH) or 2-hydroxyacetophenone (SATH) have been studied in refluxing tetrahydrofuran and complexes of types [Cp(2)TiCl(SB)]Cl (SB= SPT, SNT or
Y Atalay et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 64(1), 68-72 (2005-08-13)
The molecular geometry and vibrational frequencies of 2-amino-5-phenyl-1,3,4-thiadiazole (C8H7N3S) in the ground state has been calculated using the Hartree-Fock and density functional method (B3LYP) with 6-31G(d) basis set. The optimized geometric bond lengths and bond angles obtained by using HF
Experimental and molecular dynamics studies on corrosion inhibition of mild steel by 2-amino-5-phenyl-1, 3, 4-thiadiazole.
Tang Y, et al.
Corrosion Science, 52(1), 242-249 (2010)
Jingjing Zhang et al.
Carbohydrate polymers, 215, 108-118 (2019-04-15)
In the current study, five novel urea-functionalized chitosan derivatives were synthesized via condensation reactions of chloroacetyl chitosan (CTCS) with urea groups bearing nitrogen-containing heterocycles. In order to identify the structure characteristics of chitosan derivatives, FT-IR, 1H NMR spectroscopy, and elemental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service