Skip to Content
Merck
All Photos(1)

Key Documents

659282

Sigma-Aldrich

4-Hydroxy-3-methylbenzoic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$131.00

$131.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$131.00

About This Item

Linear Formula:
HOC6H3(CH3)CO2H
CAS Number:
Molecular Weight:
152.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$131.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

173-177 °C

functional group

carboxylic acid

SMILES string

Cc1cc(ccc1O)C(O)=O

InChI

1S/C8H8O3/c1-5-4-6(8(10)11)2-3-7(5)9/h2-4,9H,1H3,(H,10,11)

InChI key

LTFHNKUKQYVHDX-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Pan et al.
Water research, 88, 60-68 (2015-10-17)
Chlorine/chloramine residuals are maintained in drinking water distribution systems to prevent microbial contamination and microorganism regrowth. During household cooking processes (e.g., soup making), the residual chlorine/chloramines in tap water may react with the iodide in iodized table salt to form
Yang Pan et al.
Chemosphere, 144, 2312-2320 (2015-11-26)
During drinking water disinfection, iodinated disinfection byproducts (I-DBPs) can be generated through reactions between iodide, disinfectants, and natural organic matter. Drinking water I-DBPs have been increasingly attracting attention as emerging organic pollutants as a result of their significantly higher toxicity
F Guneral et al.
Clinical chemistry, 40(6), 862-866 (1994-06-01)
Organic acid concentrations were quantified by gas chromatography and the individual acids identified by mass spectrometry in urine specimens from a healthy Turkish pediatric population of ages 2 days to 16 years, subdivided into five age groups. We quantified 69

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service