Skip to Content
Merck
All Photos(2)

Key Documents

138126

Sigma-Aldrich

Methyl vanillate

99%

Synonym(s):

Methyl 4-hydroxy-3-methoxybenzoate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$162.00
100 G
$614.00

$162.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$162.00
100 G
$614.00

About This Item

Linear Formula:
HOC6H3(OCH3)CO2CH3
CAS Number:
Molecular Weight:
182.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$162.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

solid

bp

285-287 °C (lit.)

mp

64-67 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1ccc(O)c(OC)c1

InChI

1S/C9H10O4/c1-12-8-5-6(9(11)13-2)3-4-7(8)10/h3-5,10H,1-2H3

InChI key

BVWTXUYLKBHMOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Methyl vanillate is a volatile compound present in cloudberries (Rubus Chamaemorus.)[1].

Application

Methyl vanillate was used to investigate the volatile compounds of heather honeys from Calluna vulgaris and Erica arborea[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Revecca Theodori et al.
Journal of natural products, 69(4), 662-664 (2006-04-29)
Three linear sesquiterpene lactones, anthecotulide (1), hydroxyanthecotulide (2), and acetoxyanthecotulide (3), were isolated from the aerial parts of Anthemis auriculata. Five known flavonoids, taraxa-20(30)en-3beta-ol (4), and methyl vanillate (5) were also isolated. The structures and the relative configuration of the
T Pyysalo et al.
Zeitschrift fur Lebensmittel-Untersuchung und -Forschung, 163(1), 25-30 (1977-01-01)
The qualitative and quantitative differences between the volatiles of fresh and heated cloudberries have been evaluated and the significance of the differences calculated. 14 new cloudberry volatiles have been identified, including vanillin, methyl vanillate, acetovanillone and 3,5-dihydroxy-2-methyl-4H-pyran-4-one. During the heating
Floral origin markers of heather honeys: Erica arborea and Calluna vulgaris.
Guyot C, et al.
Food Chemistry, 64(1), 3-11 (1999)
Yuping Liu et al.
Food chemistry, 319, 126564-126564 (2020-03-13)
Zanthoxylum armatum DC Prodr. pericarp (ZAP) is an important spice because of its unique odor and taste. ZAP oil was obtained by supercritical carbon dioxide extraction. To characterize potent odorants in ZAP oil, volatiles were isolated by headspace solid-phase microextraction
Delphine Lamoral-Theys et al.
Bioorganic & medicinal chemistry, 18(11), 3823-3833 (2010-05-15)
A series of 33 novel divanillates and trivanillates were synthesized and found to possess promising cytostatic rather than cytotoxic properties. Several compounds under study decreased by >50% the activity of Aurora A, B, and C, and WEE1 kinase activity at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service