658243
Lithium pyrrolidinoborohydride solution
1 M in THF
Synonym(s):
Lithium trihydro-1-pyrrolidinylborate
About This Item
Recommended Products
reaction suitability
reagent type: reductant
concentration
1 M in THF
refractive index
n20/D 1.425
density
0.890 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
[Li+].[H][B-]([H])([H])N1CCCC1
InChI
1S/C4H11BN.Li/c5-6-3-1-2-4-6;/h1-4H2,5H3;/q-1;+1
InChI key
CUBZYJJJUFGOBJ-UHFFFAOYSA-N
Application
- Reducing agent for reduction of carbonic and carboxylic acid derivatives
- Reagent for reduction-amination reactions
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
1.4 °F - closed cup
Flash Point(C)
-17.0 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Lithium aminoborohydride (LAB) reagents are a new class of powerful and selective reagents developed in the laboratory of Professor Bakthan Singaram at the University of California, Santa Cruz.
Related Content
New asymmetric methodology meets the continuous demand for optically active compounds in various fields.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service