Skip to Content
Merck
All Photos(2)

Documents

653608

Sigma-Aldrich

Aluminum

powder, <5 μm particle size, 99.5% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.5% trace metals basis

form

powder

autoignition temp.

1400 °F

resistivity

2.6548 μΩ-cm

particle size

<5 μm

bp

2460 °C (lit.)

mp

660.37 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

Application

Aluminum powder can be used for a variety of applications, such as:
  • generation of hydrogen from water
  • precursor in the chemical industry
  • mechanical alloying
  • energy materials

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Sol. 1 - Water-react 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

Mechanical alloying of multi-walled carbon nanotubes and aluminium powders for the preparation of carbon/metal composites
Wang L, et al.
Carbon, 47(15), 3427-3433 (2009)
Application of activated aluminum powder for generation of hydrogen from water
Rosenband V and Gany A
International Journal of Hydrogen Energy, 35(20), 10898-10904 (2010)
Detonation properties of explosives containing nanometric aluminum powder
Proceedings of the 12th international detonation symposium, 47(15), 11-21 (2002)
Development and applications of ultrafine aluminium powders
Kearns M
Materials Science & Engineering. A, Structural Materials : Properties, Microstructure and Processing, 375(20), 120-126 (2004)
Thierry Deschamps et al.
Optics express, 21(7), 8382-8392 (2013-04-11)
Using a combination of experimental techniques such as optical absorption, Raman scattering, continuous wave and pulse Electron Spin Resonance (ESR), we characterize a set of γ-irradiated Yb(3+) doped silica glass preforms with different contents of phosphorous and aluminum. We demonstrate

Articles

Mechanical alloying is a “brute force” method of affecting alloying and chemical reactions. The mixture of reactant powders and several balls are placed in the milling jar of a high-energy ball mill, for example, a shaker mill or a planetary mill.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service