Skip to Content
Merck
All Photos(2)

Key Documents

GF82061670

Aluminum

powder, 15 max. part. size (micron), weight 200 g, purity 99%

Synonym(s):

AL006038

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$560.00

$560.00


Estimated to ship onMarch 28, 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$560.00

About This Item

Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
MDL number:
UNSPSC Code:
12141702
PubChem Substance ID:
NACRES:
NA.23

$560.00


Estimated to ship onMarch 28, 2025


Request a Bulk Order

Assay

99%

form

powder

autoignition temp.

1400 °F

manufacturer/tradename

Goodfellow 820-616-70

resistivity

2.6548 μΩ-cm

weight

200 g

max. part. size

15 (micron)

bp

2460 °C (lit.)

mp

660.37 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Pyr. Sol. 1 - Water-react 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service