Skip to Content
Merck
All Photos(1)

Key Documents

649066

Sigma-Aldrich

Chlorodicyclopentylphosphine

97%

Synonym(s):

Dicyclopentylchlorophosphine, Dicyclopentylphosphine chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$342.00

$342.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$342.00

About This Item

Empirical Formula (Hill Notation):
C10H18ClP
CAS Number:
Molecular Weight:
204.68
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$342.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

density

1.069 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

ClP(C1CCCC1)C2CCCC2

InChI

1S/C10H18ClP/c11-12(9-5-1-2-6-9)10-7-3-4-8-10/h9-10H,1-8H2

InChI key

QTYRJBRBODDGCQ-UHFFFAOYSA-N

General description

Chlorodicyclopentylphosphine is a phosphine precursor for ligand preparation in Negishi and Suzuki coupling reactions.
http://citeseerx.ist.psu.edu/viewdoc/download?doi=10.1.1.259.5096&rep=rep1&type=pdf

Application

Chlorodicyclopentylphosphine may be used to prepare:
  • Cp,Cp-MannOP, a chiral diphosphinite ligand by reacting with mannitol.[Cp=cyclopentyl; Mann=mannitol][1]
  • (S)-Cp,Cp-oxoProNOP, a bis(dicycloalkylphosphino) amidophosphine-phosphinite ligand by reacting with (S)-5-(hydroxymethyl)-2-pyrrolidinone[2]
  • LcPeNiCl, a precursor for preparing LcPeNiH, which is an efficient catalyst for the reduction of CO2[cPe=cyclopentyl][3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amidophosphine-Phosphinites: Synthesis and Use in Rhodium-Based Asymmetric Hydrogenation of Activated Keto Compounds. Crystal Structure of Bis [(?-chloro)((S)-2-((diphenylphosphino) oxy)-2-phenyl-N-(diphenylphosphino)-N-methylacetamide) rhodium (I)].
Roucoux A, et al.
Organometallics, 15(10), 2440-2449 (1996)
Catalytic properties of nickel bis (phosphinite) pincer complexes in the reduction of CO 2 to methanol derivatives.
Chakraborty S, et al.
Polyhedron, 32(1), 30-34 (2012)
New diphosphinite ligands derived from mannitol for rhodium catalyzed enantioselective hydrogenation of ketones.
Naili S, et al.
Tetrahedron Letters, 41(16), 2867-2870 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service