Skip to Content
Merck
All Photos(1)

Key Documents

643882

Sigma-Aldrich

3,5-Dimethylisoxazole-4-boronic acid pinacol ester

97%

Synonym(s):

3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl)isoxazole, 3,5-Dimethylisoxazoleboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$167.00

$167.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$167.00

About This Item

Empirical Formula (Hill Notation):
C11H18BNO3
CAS Number:
Molecular Weight:
223.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$167.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

95-99 °C (lit.)

SMILES string

Cc1noc(C)c1B2OC(C)(C)C(C)(C)O2

InChI

1S/C11H18BNO3/c1-7-9(8(2)14-13-7)12-15-10(3,4)11(5,6)16-12/h1-6H3

InChI key

CVLHETBAROWASE-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service