Skip to Content
Merck
All Photos(3)

Key Documents

641642

Sigma-Aldrich

1-Piperazinepropanol

97%

Synonym(s):

1-(3-Hydroxy-1-propyl)piperazine, 1-(3-Hydroxypropyl)-4-piperazine, 1-(3-Hydroxypropyl)piperazine, 3-(1-Piperazinyl)-1-propanol, 3-(1-Piperazinyl)propanol, 4-(3-Hydroxypropyl)piperazine, N-(3-Hydroxypropyl)piperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16N2O
CAS Number:
Molecular Weight:
144.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

49-53 °C (lit.)

functional group

hydroxyl

SMILES string

OCCCN1CCNCC1

InChI

1S/C7H16N2O/c10-7-1-4-9-5-2-8-3-6-9/h8,10H,1-7H2

InChI key

LWEOFVINMVZGAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of biologically active molecules:
  • Preparation of chloropyridazine piperazines as human rhinovirus (HRV-3) inhibitors
  • Synthesis of arylbenzoylpyrrolidine derivatives and their Hsp90 inhibitory activity
  • Synthesis of pyrrolothienopyrazines as novel serotonin-4 receptors antagonists
  • Preparation of carbamate-functionalized 2,6-disubstituted N-(arylsulfonyl) piperidines as conformationally restricted and orally active γ-secretase inhibitors

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service