Skip to Content
Merck
All Photos(1)

Key Documents

641375

Sigma-Aldrich

4-Fluorophthalic anhydride

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$141.00
25 G
$552.00

$141.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$141.00
25 G
$552.00

About This Item

Empirical Formula (Hill Notation):
C8H3FO3
CAS Number:
Molecular Weight:
166.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$141.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

75-79 °C (lit.)

functional group

anhydride
ester
fluoro

SMILES string

Fc1ccc2C(=O)OC(=O)c2c1

InChI

1S/C8H3FO3/c9-4-1-2-5-6(3-4)8(11)12-7(5)10/h1-3H

InChI key

XVMKZAAFVWXIII-UHFFFAOYSA-N

General description

4-Fluorophthalic anhydride is a fluorinated building block commonly used in chemical synthesis for the preparation of polyimide, pesticides, herbicides, and fungicides. It can be synthesized by treating 4-chlorophthalic anhydride with potassium fluoride.[1]

Application

4-Fluorophthalic anhydride can be used as a reactant to prepare:
  • 4-amino substituted phthalimides applicable as potential fluorescent probes and fluorescent bioactive compounds.[2]
  • 4,4′-bis(4-fluorophthalimido)diphenyl ether monomer, which is used in the synthesis of cardo polymers via nucleophilic substitution polymerization reaction.[3]
  • Asymmetric polyimides by a solution polycondensation reaction.[4]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of high performance phenolphthalein-based cardo poly (ether ketone imide) s via aromatic nucleophilic substitution polymerization
Liu Jiangtao, et al.
Polymer, 70, 30-37 (2015)
Efficient one-step synthesis of 4-amino substituted phthalimides and evaluation of their potential as fluorescent probes
Kindahl T and Chorell E
Organic & Biomolecular Chemistry, 12(25), 4461-4470 (2014)
Synthesis of cardo containing asymmetric poly (ether-naphthalimide-phthalimide) s
Mushtaq N, et al.
Polymer International, 66(11), 1633-1639 (2017)
Vu H Nguyen et al.
EJNMMI research, 3(1), 80-80 (2013-12-18)
Sigma2 (σ2) receptors are highly expressed in cancer cell lines and in tumours. Two novel selective 18F-phthalimido σ2 ligands, 18F-SIG343 and 18F-SIG353, were prepared and characterised for their potential tumour imaging properties. Preparation of 18F-SIG343 and 18F-SIG353 was achieved via
Saturated vapor pressure of 4-fluorophthalic anhydride and 4-chlorophthalic anhydride and isobaric vapor-liquid phase equilibrium of 4-fluorophthalic anhydride+ 4-chlorophthalic anhydride: Measurement and modeling
LiRongRong, et al.
Fluid Phase Equilibria, 396, 35-42 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service