Skip to Content
Merck
All Photos(1)

Key Documents

640611

Sigma-Aldrich

3-Chloro-7-hydroxy-4-methylcoumarin

97%

Synonym(s):

3-Chloro-4-methylumbelliferone

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$80.50
5 G
$161.00

$80.50


Estimated to ship onApril 14, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$80.50
5 G
$161.00

About This Item

Empirical Formula (Hill Notation):
C10H7ClO3
CAS Number:
Molecular Weight:
210.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$80.50


Estimated to ship onApril 14, 2025


Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

240-244 °C (lit.)

functional group

chloro
ester

SMILES string

CC1=C(Cl)C(=O)Oc2cc(O)ccc12

InChI

1S/C10H7ClO3/c1-5-7-3-2-6(12)4-8(7)14-10(13)9(5)11/h2-4,12H,1H3

InChI key

ODZHLDRQCZXQFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Simeon-Rudolf et al.
Chemico-biological interactions, 119-120, 119-128 (1999-07-27)
Inhibition of recombinant mouse wild type AChE (EC 3.1.1.7) and BChE (EC 3.1.1.8), and AChE peripheral site-directed mutants and human serum BChE variants by 4,4'-bipyridine (4,4'-BP) and the coumarin derivative 3-chloro-7-hydroxy-4-methylcoumarin (CHMC) was studied. The enzyme activity was measured with
J K Malik et al.
Toxicology letters, 7(6), 449-452 (1981-03-01)
Administration of [14C]chlorferron in a single oral dose of 0.5 and 20 mg/kg body weight to female rats resulted in a urinary excretion of greater than 74% of the given dose during the first 24 h. Approx. 8% of the
J K Malik et al.
Archives of toxicology, 48(1), 51-59 (1981-08-01)
The biotransformation and disposition of 3-chloro-4-methyl-(4-14C)-7-hydroxycoumarin [(14C) chlorferron] were investigated in rats after single oral administration. Following administration of (14C) chlorferron at 0.5 and 20 mg/kg body weight to male rats, greater than 90% of the given dose was eliminated
Jiyeon Ha et al.
Bioresource technology, 98(10), 1916-1923 (2006-09-22)
Chlorferon and diethylthiophosphate (DETP) are the hydrolysis products of coumaphos, an organophosphate pesticide. In this research, two consortia of bacterial cultures, one responsible for degrading chlorferon and the other for degrading DETP, were selectively enriched from waste cattle dip solution.
J K Malik et al.
Advances in experimental medicine and biology, 136 Pt A, 839-846 (1981-01-01)
The distribution, excretion and biotransformation of 3-chloro-4-methyl-(4-14C)-7-hydroxycoumarin [(14C) chlorferron] were studied in the rats after single oral administration. Male and female Wistar rats were given single oral doses of 0.5 and 20.0 mg/kg body weight of (14C) chlorferron and routes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service