Skip to Content
Merck
All Photos(2)

Key Documents

246883

Sigma-Aldrich

6,13-Pentacenequinone

99%

Synonym(s):

6,13-Pentacenedione

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$140.00

$140.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$140.00

About This Item

Empirical Formula (Hill Notation):
C22H12O2
CAS Number:
Molecular Weight:
308.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$140.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

solid

functional group

ketone

SMILES string

O=C1c2cc3ccccc3cc2C(=O)c4cc5ccccc5cc14

InChI

1S/C22H12O2/c23-21-17-9-13-5-1-2-6-14(13)10-18(17)22(24)20-12-16-8-4-3-7-15(16)11-19(20)21/h1-12H

InChI key

UFCVADNIXDUEFZ-UHFFFAOYSA-N

General description

Electronic structure of the lowest excited triplet state of 6,13-pentacenequinone has been studied by continuous-wave time-resolved electron paramagnetic resonance and pulsed electron nuclear double resonance[1].

Application

6,13-Pentacenequinone has been used in:
  • deposition of 6,13-pentacenequinone thin films on n-Si substrates by thermal evaporation[2]
  • as precursor to diarylpentacenes, potential organic field-effect transitors[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 63, 9699-9699 (2007)
Takuji Shimokage et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 58(6), 1201-1208 (2002-05-08)
Continuous-wave time-resolved EPR (cw-TREPR) and pulsed electron nuclear double resonance (ENDOR) studies have been carried out to clarify the electronic structure of the lowest excited triplet (Tl) state of 5,12-naphthacenequinone (5,12-NpQ) as well as 1,4-anthraquinone (1,4-AQ) and 6,13-pentacenequinone (6,13-PeQ). The
Structural and optical properties of 6, 13-pentacenequinone thin films.
Hwang DK, et al.
Applied Physics Letters, 85(23), 5568-5570 (2004)

Articles

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service