Skip to Content
Merck
All Photos(1)

Key Documents

241628

Sigma-Aldrich

1,2-Bis(2-chloroethoxy)ethane

97%

Synonym(s):

Dichlorotriethylene dioxide, Tri(ethylene glycol) dichloride, ‘Triglycol dichloride’

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$94.60
500 G
$331.00

$94.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$94.60
500 G
$331.00

About This Item

Linear Formula:
ClCH2CH2OCH2CH2OCH2CH2Cl
CAS Number:
Molecular Weight:
187.06
Beilstein:
506177
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$94.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.461 (lit.)

bp

235 °C (lit.)

density

1.197 g/mL at 25 °C (lit.)

functional group

chloro
ether

SMILES string

ClCCOCCOCCCl

InChI

1S/C6H12Cl2O2/c7-1-3-9-5-6-10-4-2-8/h1-6H2

InChI key

AGYUOJIYYGGHKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2-bis(2-chloroethoxy)ethane was used in the synthesis of 1,1′-[1,2-ethanediylbis(oxy-1,2-ethanediyl)]bis[3-methyl-1H-imidazolium-1-yl] chloride[1].

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mercury (II) partitioning from aqueous solutions with a new, hydrophobic ethylene-glycol functionalized bis-imidazolium ionic liquid.
Green Chemistry, 5(2), 129-135 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service