Skip to Content
Merck
All Photos(1)

Key Documents

637769

Sigma-Aldrich

4-Iodobiphenyl

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$69.60

$69.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$69.60

About This Item

Linear Formula:
C6H5C6H4I
CAS Number:
Molecular Weight:
280.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$69.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

110-114 °C

functional group

iodo
phenyl

SMILES string

Ic1ccc(cc1)-c2ccccc2

InChI

1S/C12H9I/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9H

InChI key

NXYICUMSYKIABQ-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Sinsheimer et al.
Journal of pharmaceutical sciences, 70(5), 546-549 (1981-05-01)
[125I]-4-Iodobiphenyl has been used as a model compound to investigate the in vivo metabolism of iodinated aromatic compounds in the rat. Material balance studies showed that 57.6% of the injected dose was excreted via feces and 41.2% via urine. Distribution
A Parkinson et al.
Biochemical pharmacology, 31(10), 1849-1856 (1982-05-15)
The in vitro metabolism of biphenyl, 4-fluoro-, 4-chloro-, 4-bromo- and 4-iodobiphenyl by cytochrome P-450-dependent monooxygenases was investigated using hepatic microsomes from immature male rats pretreated with phenobarbitone or 3-methylcholanthrene. The major route of metabolism of biphenyl and the 4-halobiphenyls was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service