Skip to Content
Merck
All Photos(1)

Key Documents

529982

Sigma-Aldrich

2-Iodobiphenyl

96%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$361.00

$361.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$361.00

About This Item

Linear Formula:
C6H5C6H4l
CAS Number:
Molecular Weight:
280.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$361.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

96%

refractive index

n20/D 1.662 (lit.)

bp

300 °C (lit.)

density

1.59 g/mL at 25 °C (lit.)

functional group

iodo
phenyl

SMILES string

Ic1ccccc1-c2ccccc2

InChI

1S/C12H9I/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9H

InChI key

QFUYDAGNUJWBSM-UHFFFAOYSA-N

General description

2-Iodobiphenyl is an ortho-halogenated biphenyl that can be synthesized from 2-aminobiphenyl.[1]

Application

2-Iodobiphenyl may be used in the synthesis of dibenziodolium tosylate by reacting with hydroxy(tosyloxy)iodo] benzene in acetonitrile.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dicyclic Hydrocarbons. I. 2-Aklylbiphenyls.
Goodman IA and Wise PH.
Journal of the American Chemical Society, 72(7), 3076-3079 (1950)
[Hydroxy (tosyloxy) iodo] benzene, a versatile reagent for the mild oxidation of aryl iodides at the iodine atom by ligand transfer.
Koser GF and Wettach RH.
The Journal of Organic Chemistry, 45(8), 1542-1543 (1980)
Michael Barclay et al.
Physical chemistry chemical physics : PCCP, 21(8), 4556-4567 (2019-02-12)
We present a combined theoretical and experimental study on the ionization and primary fragmentation channels of the mono-halogenated biphenyls; 2-chlorobiphenyl, 2-bromobiphenyl and 2-iodobiphenyl. The ionization energies (IEs) of the 2-halobiphenyls and the appearance energies (AEs) of the principal fragments are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service