Skip to Content
Merck
All Photos(1)

Key Documents

616656

Sigma-Aldrich

Betaine-(trimethyl-d9) hydrochloride

98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$3,640.00

$3,640.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$3,640.00

About This Item

Linear Formula:
(CD3)3N(Cl)CH2CO2H
Molecular Weight:
162.66
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

$3,640.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Assay

99% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

241-242 °C (lit.)

mass shift

M+9

SMILES string

Cl[H].[2H]C([2H])([2H])[N+](CC([O-])=O)(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C5H11NO2.ClH/c1-6(2,3)4-5(7)8;/h4H2,1-3H3;1H/i1D3,2D3,3D3;

InChI key

HOPSCVCBEOCPJZ-KYRNGWDOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service