Skip to Content
Merck
All Photos(1)

Key Documents

616478

Sigma-Aldrich

2,6-Dinitrotoluene-α,α,α-d3

98 atom % D, 98% (CP)

Synonym(s):

2,6-Dinitrotoluene-(methyl-d3)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$2,530.00

$2,530.00


Estimated to ship onMay 26, 2025


Request more information

Select a Size

Change View
100 MG
$2,530.00

About This Item

Empirical Formula (Hill Notation):
C7D3H3N2O4
CAS Number:
Molecular Weight:
185.15
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$2,530.00


Estimated to ship onMay 26, 2025


Request more information

isotopic purity

98 atom % D

Assay

98% (CP)

form

solid

mp

56-61 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])c1c(cccc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C7H6N2O4/c1-5-6(8(10)11)3-2-4-7(5)9(12)13/h2-4H,1H3/i1D3

InChI key

XTRDKALNCIHHNI-FIBGUPNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service