Skip to Content
Merck
All Photos(1)

Key Documents

492779

Sigma-Aldrich

Hydrazine-15N2 dihydrochloride

98 atom % 15N, 99% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$2,270.00

$2,270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
$2,270.00

About This Item

Linear Formula:
H215N15NH2 · 2HCl
CAS Number:
Molecular Weight:
106.95
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$2,270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % 15N

Assay

99% (CP)

form

solid

mp

200 °C (dec.) (lit.)

mass shift

M+2

SMILES string

Cl.Cl.[15NH2][15NH2]

InChI

1S/2ClH.H4N2/c;;1-2/h2*1H;1-2H2/i;;1+1,2+1

InChI key

LIAWOTKNAVAKCX-ZBUXRKNLSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehran Sharifi et al.
Journal of chromatographic science, 58(2), 83-90 (2019-11-12)
A new isotope dilution liquid chromatography/tandem mass spectrometric method was developed for the analysis of potential hydrazine present in tobacco smoke. The sample preparation was performed via an optimized derivatization method using an aqueous buffer:methanol solution of 2-nitrobenzaldehyde (10 g/L) used

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service