Skip to Content
Merck
All Photos(1)

Key Documents

615560

Sigma-Aldrich

1,4-Diamino(butane-d8) dihydrochloride

98 atom % D

Synonym(s):

1,4-Butane-d8-diamine dihydrochloride, Putrescine-d8 dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CD2)4NH2 · 2HCl
CAS Number:
Molecular Weight:
169.12
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

Assay

98% (CP)

form

solid

mp

280 °C (dec.) (lit.)

mass shift

M+8

SMILES string

Cl.Cl.[2H]C([2H])(N)C([2H])([2H])C([2H])([2H])C([2H])([2H])N

InChI

1S/C4H12N2.2ClH/c5-3-1-2-4-6;;/h1-6H2;2*1H/i1D2,2D2,3D2,4D2;;

InChI key

XXWCODXIQWIHQN-VHGLFXLXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guido Pelletti et al.
Forensic science international, 297, 221-227 (2019-03-05)
Among the several techniques proposed for the estimation of the Post Mortem Interval (PMI), the analysis of odorous amines has been applied in the past, with conflicting results. The aims of this study are: (i) to develop and validate a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service