Skip to Content
Merck
All Photos(1)

Key Documents

614009

Sigma-Aldrich

Hydrazine-d4 monodeuterate

98 atom % D

Synonym(s):

Hydrazine-d4 deuterate

Sign Into View Organizational & Contract Pricing

Select a Size

20 G
$2,190.00

$2,190.00


Estimated to ship onMay 26, 2025


Request more information

Select a Size

Change View
20 G
$2,190.00

About This Item

Linear Formula:
ND2ND2 · D2O
CAS Number:
Molecular Weight:
56.10
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$2,190.00


Estimated to ship onMay 26, 2025


Request more information

isotopic purity

98 atom % D

refractive index

n20/D 1.428

bp

120.1 °C (lit.)

mp

-51.7 °C (lit.)

density

1.156 g/mL at 25 °C

mass shift

M+6

storage temp.

2-8°C

SMILES string

[2H]O[2H].[2H]N([2H])N([2H])[2H]

InChI

1S/H4N2.H2O/c1-2;/h1-2H2;1H2/i/hD6

InChI key

IKDUDTNKRLTJSI-YIKVAAQNSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

165.2 °F

Flash Point(C)

74 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service