Skip to Content
Merck
All Photos(1)

Key Documents

609978

Sigma-Aldrich

Hydrogen peroxide-18O2 solution

2-3% in H2O, 90 atom % 18O

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$1,590.00
1 G
$8,020.00

$1,590.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
$1,590.00
1 G
$8,020.00

About This Item

Linear Formula:
H218O2
Molecular Weight:
38.01
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.12

$1,590.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

90 atom % 18O

Quality Level

concentration

2-3% in H2O

mass shift

M+4

storage temp.

−20°C

SMILES string

[18OH][18OH]

InChI

1S/H2O2/c1-2/h1-2H/i1+2,2+2

InChI key

MHAJPDPJQMAIIY-XPULMUKRSA-N

General description

Hydrogen peroxide-18O2, isotopically labeled analogue of hydrogen peroxide.

Application

18O-labeled H2O2 is used to determine absolute rate and pathway of peroxide formation and decay processes in surface waters of a coastal marine site and also use to measure absolute rates of dark H2O2 production in freshwater systems. It is used to study the acetylcoenzyme A (Ac-CoA) metabolism and potential therapeutic approach for cognitive disabilities on D(16) mouse model of down syndrome.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Andrew W Vermilyea et al.
Environmental science & technology, 44(8), 3066-3072 (2010-03-25)
Photochemical production is usually considered to be the main source of H2O2 in freshwater systems; here we show that significant dark production also occurs. We used isotope-labeled H2O2 as a tracer to simultaneously determine H2O2 production and decay rates in
Xiaojing Liu et al.
Cell, 175(2), 502-513 (2018-09-25)
Acetate is a major nutrient that supports acetyl-coenzyme A (Ac-CoA) metabolism and thus lipogenesis and protein acetylation. However, its source is unclear. Here, we report that pyruvate, the end product of glycolysis and key node in central carbon metabolism, quantitatively generates acetate
Bruno Pinto et al.
Neuron, 108(5), 887-904 (2020-10-08)
Microglia are brain-resident immune cells and regulate mechanisms essential for cognitive functions. Down syndrome (DS), the most frequent cause of genetic intellectual disability, is caused by a supernumerary chromosome 21, containing also genes related to the immune system. In the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service