Skip to Content
Merck
All Photos(1)

Key Documents

329878

Sigma-Aldrich

Water-18O

(for PET), 97 atom % 18O

Synonym(s):

18O Water

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$356.00
1 G
$1,070.00
20 G
$10,850.00
50 G
$21,680.00

$356.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
250 MG
$356.00
1 G
$1,070.00
20 G
$10,850.00
50 G
$21,680.00

About This Item

Linear Formula:
H218O
CAS Number:
Molecular Weight:
20.02
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

$356.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

description

Normalized with respect to hydrogen

Quality Level

isotopic purity

97 atom % 18O

Assay

≥99%

form

liquid

technique(s)

PET imaging: suitable

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.11 g/mL at 20 °C (lit.)

compatibility

(for PET)

mass shift

M+2

storage temp.

room temp

SMILES string

[18OH2]

InChI

1S/H2O/h1H2/i1+2

InChI key

XLYOFNOQVPJJNP-NJFSPNSNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Water-18O is an isotopic analogue of water.

Application

Water-18O is an isotopically modified water, which is appropriate for PET applications.
It can also be used:
  • In paleoclimatology, hydrology, and plant ecology to determine soil stable oxygen isotope analysis of plant cellulose using δ18O.[1]
  • To determine the diffusion of moisture into crystals using secondary ion mass spectrometry (SIMS).[2]

Packaging

Packaged in septum-sealed serum bottles.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Berk et al.
FEBS letters, 595(2), 230-240 (2020-10-29)
Identifying the targetome of a microRNA is key for understanding its functions. Cross-linking and immunoprecipitation (CLIP) methods capture native miRNA-mRNA interactions in cells. Some of these methods yield small amounts of chimeric miRNA-mRNA sequences via ligation of 5'-phosphorylated RNAs produced
Diffusion of Water Species in Yttria-Stabilized Zirconia
Duong T, et al.
Journal of the American Chemical Society, 92(11), 2731-2737 (2009)
Separating soil and leaf water 18O isotopic signals in plant stem cellulose
Sternberg, Leonel da Silveira Lobo and Anderson, William T and Morrison, Kanema
Geochimica et Cosmochimica Acta, 67(14), 2561-2566 (2003)
Xueyan Cao et al.
Food chemistry, 276, 266-273 (2018-11-10)
Milk glycoproteins are crucial nutrients with a variety of functions. However, whey N-glycoproteomes in human and bovine milks have not been characterized during lactation. Herein, using lectin enrichment and liquid chromatography tandem mass spectrometry, 68, 58, 100, and 98 N-glycoproteins
Elizabeth A Rettedal et al.
MicrobiologyOpen, 4(2), 208-219 (2015-02-05)
Current limitations in culture-based methods have lead to a reliance on culture-independent approaches, based principally on the comparative analysis of primary semantides such as ribosomal gene sequences. DNA can be remarkably stable in some environments, so its presence does not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service