Skip to Content
Merck
All Photos(1)

Documents

609358

Sigma-Aldrich

Potassium cyanate-15N

≥98 atom % 15N, ≥95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOC15N
CAS Number:
Molecular Weight:
82.11
MDL number:
UNSPSC Code:
12352118
PubChem Substance ID:

isotopic purity

≥98 atom % 15N

Assay

≥95% (CP)

form

solid

mass shift

M+1

SMILES string

[K+].[O-]C#[15N]

InChI

1S/CHNO.K/c2-1-3;/h3H;/q;+1/p-1/i2+1;

InChI key

GKKCIDNWFBPDBW-CGOMOMTCSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Zambonin et al.
Journal of neuroscience methods, 192(1), 115-120 (2010-07-28)
Mitochondrial defects have been implicated in the degeneration of axons in a number of CNS disorders, including multiple sclerosis. Uniquely, mitochondria harbor the only non-nuclear DNA (mitochondrial DNA or mtDNA), which encodes functionally important subunits of the respiratory chain. The
Grégoire Danger et al.
Amino acids, 42(6), 2331-2341 (2011-07-20)
The reaction of cyanate with C-terminal carboxyl groups of peptides in aqueous solution was considered as a potential pathway for the abiotic formation of peptide bonds under the condition of the primitive Earth. The catalytic effect of dicarboxylic acids on
Jirí Verner et al.
Molecules (Basel, Switzerland), 11(1), 34-42 (2007-10-27)
The reactivity of alicyclic ketazines in criss-cross cycloadditions was investigated. They react with potassium cyanate and ammonium thiocyanate in the presence of acetic acid to form spirocyclic perhydro[1,2,4]triazolo[1,2-alpha][1,2,4]triazole-1,5-diones and perhydro[1,2,4]triazolo[1,2-alpha][1,2,4]triazole-1,5-dithiones, respectively, in relatively high yields.
G Mistrello et al.
International archives of allergy and immunology, 145(3), 224-230 (2007-10-05)
Date palm (DP) pollen can cause allergic symptoms in people living in different countries. Specific immunotherapy with allergenic extracts by subcutaneous route is effective to cure allergic people. However, the risk of side effects has led to explore safer therapeutic
Mohammad Ali Ghaffari et al.
Scandinavian journal of clinical and laboratory investigation, 70(2), 122-127 (2010-02-17)
Previous studies have shown that the increase of carbamylated LDL (cLDL), a product of nonenzymatic modification of LDL in human serum by urea-derived cyanate, may cause cardiovascular complications in patients with chronic renal insufficiency. This study examined the inhibitory effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service