Skip to Content
Merck
All Photos(1)

Key Documents

490539

Sigma-Aldrich

Potassium cyanide-13C,15N

99 atom % 13C, 98 atom % 15N

Synonym(s):

13C,15N Labeled KCN, 13C,15N Labeled potassium cyanide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,890.00
500 MG
$2,560.00

$1,890.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$1,890.00
500 MG
$2,560.00

About This Item

Linear Formula:
K13C15N
CAS Number:
Molecular Weight:
67.10
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

$1,890.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C
98 atom % 15N

Quality Level

form

powder

mp

>300 °C (lit.)

mass shift

M+2

storage temp.

room temp

SMILES string

[K][13C]#[15N]

InChI

1S/CN.K/c1-2;/i1+1,2+1;

InChI key

YUZRZFQHUCKACF-AWQJXPNKSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service