Skip to Content
Merck
All Photos(1)

Key Documents

604119

Sigma-Aldrich

Iodoacetic acid-1-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$3,950.00

$3,950.00


Estimated to ship onApril 13, 2025


Request more information

Select a Size

Change View
500 MG
$3,950.00

About This Item

Linear Formula:
ICH213CO2H
CAS Number:
Molecular Weight:
186.94
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$3,950.00


Estimated to ship onApril 13, 2025


Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

form

solid

mp

77-79 °C (lit.)

mass shift

M+1

storage temp.

−20°C

SMILES string

O[13C](=O)CI

InChI

1S/C2H3IO2/c3-1-2(4)5/h1H2,(H,4,5)/i2+1

InChI key

JDNTWHVOXJZDSN-VQEHIDDOSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service