Skip to Content
Merck
All Photos(2)

Key Documents

721328

Sigma-Aldrich

Iodoacetamide-13C2, 2-d2

99 atom % 13C, 98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$3,280.00

$3,280.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
$3,280.00

About This Item

Linear Formula:
I13CD213CONH2
CAS Number:
Molecular Weight:
188.96
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

$3,280.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C
98 atom % D

Quality Level

Assay

≥99% (GC)

form

powder

mp

92-95 °C (lit.)

mass shift

M+4

storage temp.

2-8°C

SMILES string

[2H][13C]([2H])(I)[13C](N)=O

InChI

1S/C2H4INO/c3-1-2(4)5/h1H2,(H2,4,5)/i1+1D2,2+1

InChI key

PGLTVOMIXTUURA-HDZHEIKLSA-N

General description

Iodoacetamide-13C2, isotopically labeled analogue of 2-Iodoacetamide.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zhonghua Wang et al.
Cell reports, 24(4), 815-823 (2018-07-26)
SAMHD1 is a dNTP triphosphohydrolase (dNTPase) that impairs retroviral replication in a subset of non-cycling immune cells. Here we show that SAMHD1 is a redox-sensitive enzyme and identify three redox-active cysteines within the protein: C341, C350, and C522. The three
Jessica N Spradlin et al.
Nature chemical biology, 15(7), 747-755 (2019-06-19)
Nimbolide, a terpenoid natural product derived from the Neem tree, impairs cancer pathogenicity; however, the direct targets and mechanisms by which nimbolide exerts its effects are poorly understood. Here, we used activity-based protein profiling (ABPP) chemoproteomic platforms to discover that
Charles A Berdan et al.
Cell chemical biology, 26(7), 1027-1035 (2019-05-14)
Parthenolide, a natural product from the feverfew plant and member of the large family of sesquiterpene lactones, exerts multiple biological and therapeutic activities including anti-inflammatory and anti-cancer effects. Here, we further study the parthenolide mechanism of action using activity-based protein
Lydia Boike et al.
Cell chemical biology, 28(1), 4-13 (2020-09-24)
MYC is a major oncogenic transcriptional driver of most human cancers that has remained intractable to direct targeting because much of MYC is intrinsically disordered. Here, we have performed a cysteine-reactive covalent ligand screen to identify compounds that could disrupt
Mark Jelcic et al.
Cell chemical biology, 27(8), 1073-1083 (2020-06-11)
ATP is an important energy metabolite and allosteric signal in health and disease. ATP-interacting proteins, such as P2 receptors, control inflammation, cell death, migration, and wound healing. However, identification of allosteric ATP sites remains challenging, and our current inventory of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service