Skip to Content
Merck
All Photos(1)

Key Documents

602930

Sigma-Aldrich

Oxygen-16O2

99.98 atom % 16O, 18O-depleted

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
16O2
CAS Number:
Molecular Weight:
31.99
EC Number:
MDL number:
UNSPSC Code:
12141904
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99.98 atom % 16O

Assay

99% (CP)

form

gas

mass shift

depleted

SMILES string

[16O]=[16O]

InChI

1S/O2/c1-2/i1+0,2+0

InChI key

MYMOFIZGZYHOMD-ZCWHFVSRSA-N

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame over circleGas cylinder

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Gas 1 - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meiying Liu et al.
Dalton transactions (Cambridge, England : 2003), 49(31), 10895-10900 (2020-07-29)
Ferroelectrics as crucial functional materials have attracted much interest since ferroelectricity was discovered in 1920. Herein, an unusual high-frequency ferroelectric, (CH3)2NH·HCl@Cd-MOF, was successfully obtained through a dual-step synthetic methodology. A chiral porous Cd-MOF with a channel size of 6.8 ×
Jiahong Lin et al.
International journal of biological macromolecules, 84, 189-199 (2015-12-23)
Increasing attention has been given to nanobiocatalysis for commercial applications. In this study, laccase was reversibly immobilized onto Cu(ΙΙ)- and Mn(ΙΙ)-chelated magnetic microspheres and successfully applied to remove bisphenol A (BPA) from water. The results indicated that the loading of
Andrey A Zaspa et al.
Journal of bioenergetics and biomembranes, 52(6), 495-504 (2020-11-16)
The light-induced functioning of photosynthetic pigment-protein complex of photosystem II (PSII) is linked to the vectorial translocation of charges across the membrane, which results in the formation of voltage. Direct measurement of the light-induced voltage (∆V) generated by spinach oxygen-evolving
Elvira Romero et al.
ACS chemical biology, 15(2), 504-512 (2020-01-23)
The azoreductase AzoA from the alkali-tolerant Bacillus wakoensis A01 has been studied to reveal its structural and mechanistic details. For this, a recombinant expression system was developed which yields impressive amounts of fully active enzyme. The purified holo enzyme is
Marek Pająk et al.
Chemistry Central journal, 11(1), 90-90 (2017-11-01)
The paper examines Co(II)-amino acid-imidazole systems (where amino acid = L-α-amino acid: alanine, asparagine, histidine) which, when in aqueous solutions, activate and reversibly take up dioxygen, while maintaining the structural scheme of the heme group (imidazole as axial ligand and O Investigations

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service