Skip to Content
Merck
All Photos(1)

Documents

592021

Sigma-Aldrich

Nicotinamide-(amide-15N)

98 atom % 15N, 98% (CP)

Synonym(s):

3-Pyridine(carboxamide-15N), Vitamin B3-(amide-15N)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H615NNO
CAS Number:
Molecular Weight:
123.12
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % 15N

Assay

98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

color

white

mp

130-132 °C (lit.)

mass shift

M+1

SMILES string

[15NH2]C(=O)c1cccnc1

InChI

1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)/i7+1

InChI key

DFPAKSUCGFBDDF-CDYZYAPPSA-N

General description

Nicotinamide (or niacinamide) is an amide form of vitamin B3/nicotinic acid that can be used in a variety of cosmetic and pharmaceutical applications. It is found in meat, fish, nuts, and mushrooms, as well as in some vegetables. It is also a precursor for essential coenzymes.

Application

Nicotinamide-(amide-15N) can be utilized in the biosynthesis of nicotinamide adenine dinucleotide (NAD). This product can also be used as an analytical standard.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A review of nicotinamide: treatment of skin diseases and potential side effects
Rolfe HM
Journal of Cosmetic Dermatology, 13(4), 324-328 (2014)
Safety of high-dose nicotinamide: a review
Knip M, et al.
Diabetologia, 43(11), 1337-1345 (2000)
Igor Shats et al.
Cell metabolism, 31(3), 564-579 (2020-03-05)
Nicotinamide adenine dinucleotide (NAD), a cofactor for hundreds of metabolic reactions in all cell types, plays an essential role in metabolism, DNA repair, and aging. However, how NAD metabolism is impacted by the environment remains unclear. Here, we report an
Rong Grace Zhai et al.
Cellular and molecular life sciences : CMLS, 66(17), 2805-2818 (2009-05-19)
Nicotinamide/nicotinic acid mononucleotide adenylyltransferase (NMNAT) has long been known as the master enzyme in NAD biosynthesis in living organisms. A burst of investigations on NMNAT, going beyond enzymology, have paralleled increasing discoveries of key roles played by NAD homeostasis in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service