Merck
  • Nicotinamide/nicotinic acid mononucleotide adenylyltransferase, new insights into an ancient enzyme.
Nicotinamide/nicotinic acid mononucleotide adenylyltransferase, new insights into an ancient enzyme.
Cellular and molecular life sciences : CMLS (2009-05-19)
Rong Grace Zhai, Menico Rizzi, Silvia Garavaglia
ABSTRACT
Nicotinamide/nicotinic acid mononucleotide adenylyltransferase (NMNAT) has long been known as the master enzyme in NAD biosynthesis in living organisms. A burst of investigations on NMNAT, going beyond enzymology, have paralleled increasing discoveries of key roles played by NAD homeostasis in a number or patho-physiological conditions. The availability of in-depth kinetics and structural enzymology analyses carried out on NMNATs from different organisms offer a powerful tool for uncovering fascinating evolutionary relationships. On the other hand, additional functions featuring NMNAT have emerged from investigations aimed at unraveling the molecular mechanisms responsible for complex biological phenomena such as neurodegeneration. NMNAT appears to be a multifunctional protein that sits both at the core of central metabolism and at a crossroads of multiple cellular processes. The resultant wealth of biochemical data has built a robust framework upon which design of NMNAT activators, inhibitors or enzyme variants of potential medical interest can be based.
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Supelco
Nicotinamide (Niacinamide), analytical standard
Sigma-Aldrich
Nicotinamide-(amide-15N), 98 atom % 15N, 98% (CP)