Skip to Content
Merck
All Photos(1)

Key Documents

588245

Sigma-Aldrich

Bis(4-aminophenyl)ether-15N2

98 atom % 15N

Synonym(s):

4,4′-Oxydianiline-15N2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O(C6H415NH2)2
CAS Number:
Molecular Weight:
202.22
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % 15N

form

solid

bp

188-192 °C (lit.)

mass shift

M+2

SMILES string

[15NH2]c1ccc(Oc2ccc([15NH2])cc2)cc1

InChI

1S/C12H12N2O/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12/h1-8H,13-14H2/i13+1,14+1

InChI key

HLBLWEWZXPIGSM-SBAVNFSASA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

426.2 °F - closed cup

Flash Point(C)

219 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service