Skip to Content
Merck
All Photos(1)

Key Documents

541710

Sigma-Aldrich

2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$109.00

$109.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$109.00

About This Item

Linear Formula:
(F3C)2C(C6H3(NH2)OH)2
CAS Number:
Molecular Weight:
366.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$109.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

245-248 °C (lit.)

functional group

fluoro

SMILES string

Nc1cc(ccc1O)C(c2ccc(O)c(N)c2)(C(F)(F)F)C(F)(F)F

InChI

1S/C15H12F6N2O2/c16-14(17,18)13(15(19,20)21,7-1-3-11(24)9(22)5-7)8-2-4-12(25)10(23)6-8/h1-6,24-25H,22-23H2

InChI key

MSTZGVRUOMBULC-UHFFFAOYSA-N

General description

2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane, also referred to as 4,4′-(1,1,1,3,3,3-hexafluoro-2,2-propanediyl)bis(2-aminophenol), is an aromatic diamine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang-Yen Yu et al.
Polymers, 12(5) (2020-05-10)
High-transparency soluble polyimide with COOH and fluorine functional groups and TiO2-SiO2 composite inorganic nanoparticles with high dielectric constants were synthesized in this study. The polyimide and inorganic composite nanoparticles were further applied in the preparation of organic-inorganic hybrid high dielectric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service