Skip to Content
Merck
All Photos(1)

Documents

579602

Sigma-Aldrich

(R)-(+)-1,2-Dithiolane-3-pentanoic acid

97%

Synonym(s):

(R)-6,8-Dithiooctanoic acid, (R)-6,8-Thioctic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O2S2
CAS Number:
Molecular Weight:
206.33
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

form

solid

optical activity

[α]20/D +104°, c = 1 in chloroform

mp

48-52 °C (lit.)

SMILES string

OC(=O)CCCC[C@@H]1CCSS1

InChI

1S/C8H14O2S2/c9-8(10)4-2-1-3-7-5-6-11-12-7/h7H,1-6H2,(H,9,10)/t7-/m1/s1

InChI key

AGBQKNBQESQNJD-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michela Rosini et al.
Journal of medicinal chemistry, 48(2), 360-363 (2005-01-22)
The coupling of two different pharmacophores, each endowed with different biological properties, afforded the hybrid compound lipocrine (7), whose biological profile was markedly improved relative to those of prototypes tacrine and lipoic acid. Lipocrine is the first compound that inhibits
Alessandra Antonello et al.
Journal of medicinal chemistry, 48(1), 28-31 (2005-01-07)
To combine in the same molecule alpha(1)-adrenoreceptor blocking and antioxidant properties, compounds 2-5 were designed and synthesized. All compounds were effective alpha(1)-adrenoreceptor antagonists and were tested in both functional and binding assays. In addition, compounds 2 and 5 also displayed
B Steinmann et al.
Helvetica paediatrica acta, 34(6), 589-599 (1979-01-01)
Strychnine, a potent antagonist of glycine was given to twins suffering from nonketotic hyperglycinemia at age 73 hours. Within hours of the onset of treatment favorable effects were observed such as improvements of muscle tone, movements, defense reactions, and probably
T Tankova et al.
International journal of clinical practice, 59(6), 645-650 (2005-06-01)
Twenty-three diabetic patients -- 16 men and seven women (mean age: 50.7 +/- 17.4 years; mean duration of diabetes: 13.6 +/- 6.9 years) -- with diabetic mononeuropathy of the cranial nerves participated in the study. Four of them were with
Daniel Konrad
Antioxidants & redox signaling, 7(7-8), 1032-1039 (2005-07-07)
Alpha-lipoic acid is a naturally occurring cofactor of mitochondrial dehydrogenase complexes and a potent antioxidant. It can interchange between a reduced form and an oxidized form, thereby displaying reducing (antioxidant) and prooxidant properties, respectively. It is suggested that alpha-lipoic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service