Skip to Content
Merck
All Photos(2)

Key Documents

576182

Sigma-Aldrich

4(5)-(Hydroxymethyl)imidazole

97%

Synonym(s):

1H-Imidazol-4-ylmethanol, 1H-Imidazol-5-ylmethanol, 1H-Imidazole-4-methanol (9CI), 4-(Hydroxymethyl)imidazole, 4-Imidazolylmethanol, 4-Methylol-1H-imidazole, 5-(Hydroxymethyl)imidazole, Imidazole-4-methanol (8CI)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$97.30

$97.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$97.30

About This Item

Empirical Formula (Hill Notation):
C4H6N2O
CAS Number:
Molecular Weight:
98.10
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$97.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

88-92 °C (lit.)

functional group

hydroxyl

SMILES string

OCc1c[nH]cn1

InChI

1S/C4H6N2O/c7-2-4-1-5-3-6-4/h1,3,7H,2H2,(H,5,6)

InChI key

QDYTUZCWBJRHKK-UHFFFAOYSA-N

General description

4(5)-(Hydroxymethyl)imidazole can be prepared using 4(5)-(hydroxymethyl)imidazole picrate as the starting material.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

?The Preparation of 4(5)-hydroxymethylimidazole?
Darby.JW, et al.
Journal of the American Chemical Society, 64(02), 463-464 (1942)
Timothy R Fennell et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 123, 181-194 (2018-10-15)
4-Methylimidazole (4-MeI) is a widely used chemical, also identified as a by-product of heating foods. In cancer bioassays, 4-MeI induced lung tumors in mice, but not in rats. To establish if metabolic differences could explain species difference in carcinogenicity, this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service